Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nfkbil1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_010909.4 | NP_035039.1 | 2 - 3 | 449 | 148 | |
GenBank mRNA | AF095902.1 | - | 2 - 3 | 455 | 148 | |
AK133517.1 | - | 2 - 3 | 394 | 148 | ||
BC013269.1 | - | 2 - 3 | 427 | 4188066 | 148 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Probe spans exons |