Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Spon1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_145584.2 | NP_663559.1 | 11 - 12 | 1824 | 73 | |
GenBank mRNA | AK146506.1 | - | 11 - 12 | 1648 | 73 | |
BC020531.1 | - | 11 - 12 | 1694 | 4221758 | 73 | |
BC030339.1 | - | 2 - 3 | 278 | 73 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons |