Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR1800007869.mm.1 |
NM_001161483.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1800007869.mm.1 |
NM_001161483.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1800007869.mm.1 |
NM_199024.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1800007869.mm.1 |
NM_199024.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1800007869.mm.1 |
TC1800001063.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1800007873.mm.1 |
NM_199024.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1800007873.mm.1 |
NM_199024.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1800007873.mm.1 |
TC1800001063.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1800007874.mm.1 |
NM_199024.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1800007874.mm.1 |
NM_199024.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1800007874.mm.1 |
TC1800001063.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1800007875.mm.1 |
NM_199024.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1800007875.mm.1 |
NM_199024.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1800007875.mm.1 |
TC1800001063.mm.1 |
MTA TC |