Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nol4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001161483.1 | NP_001154955.1 | 4 - 5 | 480 | 94 | |
NM_199024.2 | NP_950189.2 | 4 - 5 | 2250 | 94 | ||
GenBank mRNA | AK031008.1 | - | 4 - 5 | 2250 | 94 | |
BC056377.1 | - | 4 - 5 | 473 | 6850243 | 94 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Mus musculus Probe spans exons |