Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nol4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_199024.2 | NP_950189.2 | 7 - 8 | 2851 | 71 | |
GenBank mRNA | AK029800.1 | - | 3 - 4 | 585 | 71 | |
AK031008.1 | - | 7 - 8 | 2851 | 71 | ||
AK031753.1 | - | 1 - 2 | 266 | 71 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mus musculus Probe spans exons |