Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Erbb3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_010153.1 | NP_034283.1 | 18 - 19 | 2166 | 97 | |
GenBank mRNA | AY686636.1 | - | 18 - 19 | 2166 | 97 | |
BC106091.1 | - | 2317 | 30844039 | 97 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Apoptosis Protein Kinase Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |