Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ikzf4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AK089522.1 | - | 8 - 9 | 1732 | 109 |
Set Membership: |
Amplicon length greater than or equal to 101 Mus musculus Probe spans exons |