Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rab5b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_177411.4 | NP_803130.1 | 4 - 5 | 620 | 115 | |
GenBank mRNA | AB232594.1 | - | 3 - 4 | 481 | 115 | |
AK081251.1 | - | 4 - 5 | 616 | 115 | ||
BC137574.1 | - | 3 - 4 | 532 | 8860593 | 115 | |
BC137575.1 | - | 3 - 4 | 532 | 8860594 | 115 | |
X84239.1 | - | 3 - 4 | 489 | 115 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Recommended Assay Mus musculus Inventoried Probe spans exons |