Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slc17a4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_177016.3 | NP_795990.2 | 8 - 9 | 1095 | 57 | |
GenBank mRNA | AK033669.1 | - | 8 - 9 | 1098 | 57 | |
AK162320.1 | - | 7 - 8 | 1538 | 57 | ||
BC147415.1 | - | 8 - 9 | 1056 | 9056491 | 57 | |
BC147416.1 | - | 8 - 9 | 1056 | 9056498 | 57 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Transporter Mammalian Gene Collection Mus musculus Probe spans exons |