Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Arid4b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_194262.2 | NP_919238.1 | 6 - 7 | 644 | 119 | |
NM_198122.2 | NP_937755.1 | 6 - 7 | 644 | 119 | ||
GenBank mRNA | AK031062.1 | - | 6 - 7 | 627 | 119 | |
AK041414.1 | - | 6 - 7 | 641 | 119 | ||
AK136105.1 | - | 6 - 7 | 527 | 119 | ||
AK161669.1 | - | 6 - 7 | 630 | 119 | ||
BC137783.1 | - | 5 - 6 | 406 | 8860802 | 119 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Probe spans exons |