Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Epha1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_023580.4 | NP_076069.2 | 1 - 2 | 149 | 136 | |
GenBank mRNA | AK017662.1 | - | 1 - 2 | 117 | 136 | |
AK028478.1 | - | 1 - 2 | 150 | 136 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Protein Kinase Mus musculus Probe spans exons |