Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Wrnip1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_030215.3 | NP_084491.3 | 4 - 5 | 1678 | 65 | |
GenBank mRNA | AB056151.1 | - | 4 - 5 | 1527 | 65 | |
AF208046.1 | - | 4 - 5 | 923 | 65 | ||
AK041886.1 | - | 4 - 5 | 1634 | 65 | ||
AK050368.1 | - | 4 - 5 | 1680 | 65 | ||
AK167570.1 | - | 4 - 5 | 1679 | 65 | ||
AK170542.1 | - | 4 - 5 | 1671 | 65 | ||
AK170846.1 | - | 4 - 5 | 1659 | 65 | ||
BC010482.1 | - | 4 - 5 | 1670 | 3492125 | 65 | |
BC058744.1 | - | 4 - 5 | 1068 | 65 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus Probe spans exons |