Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gmds | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_146041.2 | NP_666153.1 | 2 - 3 | 210 | 140 | |
GenBank mRNA | AK086076.1 | - | 2 - 3 | 252 | 140 | |
BC031788.1 | - | 2 - 3 | 210 | 4165509 | 140 | |
BC093502.1 | - | 2 - 3 | 234 | 4215865 | 140 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |