Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Iqub | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_172535.3 | NP_766123.2 | 8 - 9 | 1493 | 82 | |
GenBank mRNA | AK029931.1 | - | 8 - 9 | 1483 | 82 | |
AK133019.1 | - | 8 - 9 | 1475 | 82 | ||
AK161501.1 | - | 2 - 3 | 318 | 82 | ||
BC137937.1 | - | 8 - 9 | 1460 | 8860956 | 82 | |
BC137957.1 | - | 8 - 9 | 1460 | 8860976 | 82 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Recommended Assay Mus musculus Probe spans exons |