Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Zfpm2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_011766.5 | NP_035896.1 | 5 - 6 | 1032 | 76 | |
GenBank mRNA | AF118845.1 | - | 5 - 6 | 802 | 76 | |
AF125166.1 | - | 5 - 6 | 1033 | 76 | ||
BC059241.1 | - | 5 - 6 | 932 | 6408975 | 76 | |
EF514219.1 | - | 5 - 6 | 1855 | 76 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons |