Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ica1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001252266.1 | NP_001239195.1 | 10 - 11 | 1060 | 144 | |
NM_010492.3 | NP_034622.3 | 12 - 13 | 1536 | 207 | ||
GenBank mRNA | AB303567.1 | - | 11 - 12 | 1047 | 207 | |
CB056959.1 | - | 6 - 7 | 570 | 144 | ||
DV658174.1 | - | 6 - 7 | 446 | 207 | ||
U26459.1 | - | 12 - 13 | 1159 | 207 | ||
U26460.1 | - | 11 - 12 | 1065 | 207 | ||
U26461.1 | - | 10 - 11 | 1001 | 144 | ||
U34595.1 | - | 10 - 11 | 1032 | 207 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mus musculus Probe spans exons |