Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dlx5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_010056.3 | NP_034186.2 | 1 - 2 | 534 | 110 | |
GenBank mRNA | AF022075.1 | - | 1 - 2 | 373 | 110 | |
AF022076.1 | - | 1 - 2 | 373 | 110 | ||
AF033011.1 | - | 1 - 2 | 374 | 110 | ||
AF072452.1 | - | 1 - 2 | 534 | 110 | ||
AK134920.1 | - | 1 - 2 | 409 | 110 | ||
AK158772.1 | - | 1 - 2 | 469 | 110 | ||
U67840.1 | - | 2 - 3 | 510 | 110 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mus musculus 1700 Inventoried Probe spans exons |