Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fam3c | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | BC055853.1 | - | 1 - 2 | 102 | 4012164 | 100 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Mus musculus Probe spans exons |