Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dsg4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_181564.2 | NP_853543.1 | 8 - 9 | 1138 | 122 | |
GenBank mRNA | AY170623.1 | - | 1 - 2 | 86 | 122 | |
AY191584.1 | - | 8 - 9 | 1138 | 122 | ||
AY227349.1 | - | 8 - 9 | 1004 | 122 | ||
BC109009.2 | - | 8 - 9 | 1138 | 122 | ||
BC150475.2 | - | 8 - 9 | 1138 | 122 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Probe spans exons |