Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mon2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001163024.1 | NP_001156496.1 | 19 - 20 | 2903 | 122 | |
NM_001163025.1 | NP_001156497.1 | 19 - 20 | 2906 | 122 | ||
NM_153395.2 | NP_700444.2 | 19 - 20 | 2903 | 122 | ||
GenBank mRNA | AB014059.1 | - | 19 - 20 | 2425 | 122 | |
AK122425.1 | - | 19 - 20 | 2903 | 122 | ||
BC048406.1 | - | 6 - 7 | 679 | 122 | ||
BC055354.1 | - | 2 - 3 | 149 | 122 | ||
BC150943.1 | - | 19 - 20 | 2427 | 9087854 | 122 | |
CX122556.1 | - | 6 - 7 | 517 | 122 | ||
CX224591.1 | - | 4 - 5 | 297 | 122 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Probe spans exons |