Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Lmo1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AW494084.1 | - | 1 - 2 | 275 | 115 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Mus musculus Probe spans exons |