Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR0600016559.mm.1 |
TC0600002099.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0600016559.mm.1 |
NM_028421.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600016559.mm.1 |
NM_028421.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600016555.mm.1 |
TC0600002099.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0600016555.mm.1 |
NM_028421.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600016555.mm.1 |
NM_028421.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600016557.mm.1 |
TC0600002099.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0600016557.mm.1 |
NM_028421.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600016557.mm.1 |
NM_028421.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600016558.mm.1 |
TC0600002099.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0600016558.mm.1 |
NM_028421.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600016558.mm.1 |
NM_028421.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600016556.mm.1 |
TC0600002099.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0600016556.mm.1 |
NM_028421.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600016556.mm.1 |
NM_028421.1 |
Refseq |