Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cttnbp2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_080285.1 | NP_525024.1 | 6 - 7 | 2423 | 122 | |
GenBank mRNA | AK032356.1 | - | 4 - 5 | 804 | 122 | |
AK081137.1 | - | 2 - 3 | 886 | 122 | ||
AK173254.1 | - | 6 - 7 | 2329 | 122 | ||
BC068156.1 | - | 6 - 7 | 2184 | 122 | ||
BC141407.1 | - | 6 - 7 | 2419 | 9056020 | 122 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Probe spans exons |