Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Plekha7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AK040271.1 | - | 13 - 14 | 1984 | 88 | |
AK146866.1 | - | 2175 | 88 | |||
AK160810.1 | - | 13 - 14 | 2064 | 88 | ||
BC060678.1 | - | 12 - 13 | 1905 | 88 | ||
BC084587.1 | - | 13 - 14 | 2079 | 3602481 | 88 | |
EU380771.1 | - | 2201 | 88 | |||
EU380772.1 | - | 2259 | 88 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Mus musculus Probe spans exons |