Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sema5a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_009154.2 | NP_033180.2 | 16 - 17 | 2730 | 68 | |
GenBank mRNA | AK043386.1 | - | 16 - 17 | 2442 | 68 | |
AK077021.1 | - | 6 - 7 | 1184 | 68 | ||
AK141612.1 | - | 16 - 17 | 2674 | 68 | ||
AK143026.1 | - | 16 - 17 | 2741 | 68 | ||
BC065137.1 | - | 14 - 15 | 1958 | 30548165 | 68 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus Probe spans exons |