Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Abca16 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001278943.1 | NP_001265872.1 | 12 - 13 | 1915 | 134 | |
NM_001278944.1 | NP_001265873.1 | 15 - 16 | 2384 | 134 | ||
GenBank mRNA | AK161253.1 | - | 3 - 4 | 2469 | 134 | |
AY243472.1 | - | 15 - 16 | 2383 | 134 |
Set Membership: |
Amplicon length greater than or equal to 101 Transporter Mus musculus Probe spans exons |