Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Umod | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001278605.1 | NP_001265534.1 | 9 - 10 | 1975 | 61 | |
NM_009470.5 | NP_033496.1 | 9 - 10 | 2031 | 61 | ||
GenBank mRNA | AK085460.1 | - | 9 - 10 | 2025 | 61 | |
AK144065.1 | - | 9 - 10 | 1947 | 61 | ||
AK164688.1 | - | 9 - 10 | 1947 | 61 | ||
AK165507.1 | - | 9 - 10 | 1891 | 61 | ||
BC012973.1 | - | 8 - 9 | 1869 | 4210940 | 61 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus Probe spans exons |