Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Klf10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001289471.1 | NP_001276400.1 | 3 - 4 | 1295 | 106 | |
NM_013692.3 | NP_038720.1 | 3 - 4 | 1295 | 106 | ||
GenBank mRNA | AF049879.1 | - | 3 - 4 | 1291 | 106 | |
AF064088.1 | - | 3 - 4 | 1287 | 106 | ||
AK004906.1 | - | 3 - 4 | 1294 | 106 | ||
AK043433.1 | - | 3 - 4 | 1291 | 106 | ||
AK079679.1 | - | 3 - 4 | 1293 | 106 | ||
AK145676.1 | - | 3 - 4 | 1168 | 106 | ||
AK154719.1 | - | 3 - 4 | 1291 | 106 | ||
BC003316.1 | - | 3 - 4 | 1235 | 3155247 | 106 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |