Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR0600003231.mm.1 |
TC0600000416.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0600003231.mm.1 |
NM_001171003.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600003231.mm.1 |
NM_001171003.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600003233.mm.1 |
TC0600000416.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0600003233.mm.1 |
NM_001171003.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600003233.mm.1 |
NM_001171003.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600003232.mm.1 |
TC0600000416.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0600003232.mm.1 |
NM_001171003.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600003232.mm.1 |
NM_001171003.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600003230.mm.1 |
TC0600000416.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0600003230.mm.1 |
NM_001171003.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600003230.mm.1 |
NM_001171003.1 |
Refseq |