Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fam3c | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_138587.4 | NP_613053.3 | 5 - 6 | 406 | 84 | |
GenBank mRNA | AK154498.1 | - | 5 - 6 | 436 | 84 | |
AK170887.1 | - | 5 - 6 | 455 | 84 | ||
AY424275.1 | - | 5 - 6 | 406 | 84 | ||
BC009086.1 | - | 5 - 6 | 406 | 2811008 | 84 | |
BC055853.1 | - | 6 - 7 | 644 | 4012164 | 84 | |
BC069880.1 | - | 4 - 5 | 532 | 84 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons |