Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Prl7b1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_029355.2 | NP_083631.1 | 2 - 3 | 264 | 149 | |
GenBank mRNA | AF525156.1 | - | 2 - 3 | 264 | 149 | |
AK005458.1 | - | 2 - 3 | 265 | 149 | ||
BC116772.1 | - | 2 - 3 | 241 | 40126091 | 149 | |
BC119183.1 | - | 2 - 3 | 241 | 8733932 | 149 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Probe spans exons |