Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR1000010758.mm.1 |
NM_001252387.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1000010758.mm.1 |
NM_001252387.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1000010758.mm.1 |
NM_182807.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1000010758.mm.1 |
NM_182807.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1000010758.mm.1 |
NR_045514.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1000010758.mm.1 |
NR_045514.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1000010758.mm.1 |
TC1000001518.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1000010757.mm.1 |
NM_001252387.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1000010757.mm.1 |
NM_001252387.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1000010757.mm.1 |
NM_182807.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1000010757.mm.1 |
NM_182807.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1000010757.mm.1 |
NR_045514.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1000010757.mm.1 |
NR_045514.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1000010757.mm.1 |
TC1000001518.mm.1 |
MTA TC |