Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slc13a1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_019481.2 | NP_062354.2 | 13 - 14 | 1540 | 88 | |
GenBank mRNA | AF199365.1 | - | 13 - 14 | 1540 | 88 | |
AF199366.1 | - | 13 - 14 | 1540 | 88 | ||
AK005153.1 | - | 7 - 8 | 874 | 88 | ||
AK128899.1 | - | 13 - 14 | 1540 | 88 | ||
BC022672.1 | - | 9 - 10 | 919 | 88 | ||
BC040789.1 | - | 13 - 14 | 1514 | 88 | ||
BC049981.1 | - | 13 - 14 | 1522 | 5065026 | 88 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Transporter Mammalian Gene Collection Mus musculus Probe spans exons |