Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Prl7a2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_011168.4 | NP_035298.1 | 2 - 3 | 244 | 160 | |
GenBank mRNA | AF011382.1 | - | 2 - 3 | 233 | 160 | |
AF020524.1 | - | 2 - 3 | 240 | 160 | ||
AK005455.1 | - | 2 - 3 | 245 | 160 | ||
AK028068.1 | - | 2 - 3 | 244 | 160 | ||
AK131665.1 | - | 2 - 3 | 245 | 160 | ||
BC099468.1 | - | 2 - 3 | 223 | 30789902 | 160 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Probe spans exons |