Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Scnn1b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001272023.1 | NP_001258952.1 | 6 - 7 | 1145 | 81 | |
NM_011325.2 | NP_035455.1 | 6 - 7 | 1142 | 81 | ||
NR_073548.1 | 6 - 7 | 1142 | 81 | |||
GenBank mRNA | AF112186.1 | - | 6 - 7 | 1069 | 81 | |
AK028909.1 | - | 6 - 7 | 1124 | 81 | ||
AK036819.1 | - | 6 - 7 | 1143 | 81 | ||
AK087055.1 | - | 6 - 7 | 1104 | 81 | ||
AK164709.1 | - | 6 - 7 | 1105 | 81 | ||
BC131969.1 | - | 6 - 7 | 1058 | 40130246 | 81 | |
BC132397.1 | - | 6 - 7 | 1058 | 40130674 | 81 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Ion Channel Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |