Array Name |
Probe IDs |
Targets |
Target Type |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P25477113 |
- |
- |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P25477111 |
- |
- |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P25477110 |
- |
- |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P25477112 |
- |
- |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P25477108 |
- |
- |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P25477109 |
- |
- |
Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_52_P191468 |
AK140891.1 |
mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_52_P191468 |
AK077395.1 |
mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_52_P191468 |
NM_138589.2 |
mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_52_P191468 |
AK011862.1 |
mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_52_P191468 |
AK140891.1 |
mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_52_P191468 |
AK077395.1 |
mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_52_P191468 |
NM_138589.2 |
mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_52_P191468 |
AK011862.1 |
mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_55_P2108754 |
AK078990.1 |
mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_55_P2108754 |
NM_138589.2 |
mouse_transcriptome |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
831850 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
811913 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
323221 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
211124 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
894084 |
XM_001473905.1 |
refseq_mrna_mouse_XM |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
441434 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
363735 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
308410 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
71261 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
716535 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 4x180K |
153039 |
- |
- |
Agilent Mouse Genome CGH Microarray 44K G4426B |
A_53_P124329 |
- |
- |
Agilent Mouse Genome CGH Microarray 44K G4426B |
A_53_P170915 |
- |
- |
Agilent Mouse Genome CGH Microarray 44K G4426B |
A_53_P109275 |
- |
- |
SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_52_P191468 |
AK140891.1 |
mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_52_P191468 |
AK077395.1 |
mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_52_P191468 |
NM_138589.2 |
mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_52_P191468 |
AK011862.1 |
mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_52_P191468 |
AK140891.1 |
mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_52_P191468 |
AK077395.1 |
mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_52_P191468 |
NM_138589.2 |
mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_52_P191468 |
AK011862.1 |
mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_55_P2108754 |
AK078990.1 |
mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_55_P2108754 |
NM_138589.2 |
mouse_transcriptome |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P25477144 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P25477148 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P25477126 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P25477113 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P25477125 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P25477111 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P25477109 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P25477115 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P25477150 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P25477130 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P25477138 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P25477146 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P25477141 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P25477140 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P25477147 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P25477110 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P25477117 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P25477123 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P25477114 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P25477134 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P25477119 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P25477122 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P25477127 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P25477116 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P25477133 |
- |
- |
Agilent Mouse Genome CGH Microarray 244A (G4415A) |
A_53_P124329 |
- |
- |
Agilent Mouse Genome CGH Microarray 244A (G4415A) |
A_67_P01622347 |
- |
- |
Agilent Mouse Genome CGH Microarray 244A (G4415A) |
A_53_P170915 |
- |
- |
Agilent Mouse Genome CGH Microarray 244A (G4415A) |
A_53_P109275 |
- |
- |
Whole Mouse Genome G4122A |
A_52_P191468 |
AK140891.1 |
mouse_transcriptome |
Whole Mouse Genome G4122A |
A_52_P191468 |
AK077395.1 |
mouse_transcriptome |
Whole Mouse Genome G4122A |
A_52_P191468 |
NM_138589.2 |
mouse_transcriptome |
Whole Mouse Genome G4122A |
A_52_P191468 |
AK011862.1 |
mouse_transcriptome |
Whole Mouse Genome G4122A |
A_52_P191468 |
AK140891.1 |
mouse_transcriptome |
Whole Mouse Genome G4122A |
A_52_P191468 |
AK077395.1 |
mouse_transcriptome |
Whole Mouse Genome G4122A |
A_52_P191468 |
NM_138589.2 |
mouse_transcriptome |
Whole Mouse Genome G4122A |
A_52_P191468 |
AK011862.1 |
mouse_transcriptome |
Whole Mouse Genome G4122A |
A_51_P496604 |
AK140891.1 |
mouse_transcriptome |
Whole Mouse Genome G4122A |
A_51_P496604 |
NM_138589.2 |
mouse_transcriptome |
Whole Mouse Genome G4122A |
A_51_P496604 |
AK017851.1 |
mouse_transcriptome |
Whole Mouse Genome G4122A |
A_52_P88934 |
AK033428.1 |
mouse_transcriptome |
Whole Mouse Genome G4122A |
A_52_P121641 |
AK078990.1 |
mouse_transcriptome |
Whole Mouse Genome G4122A |
A_52_P121641 |
NM_138589.2 |
mouse_transcriptome |
Agilent Mouse Developmental Microarray 44K G2519A Option 002 |
A_66_P102320 |
- |
- |
Agilent Mouse Genome CGH Microarray 105A (G4416A) |
A_53_P124329 |
- |
- |
Agilent Mouse Genome CGH Microarray 105A (G4416A) |
A_53_P109275 |
- |
- |
Agilent Mouse Genome CGH Microarray 105A (G4416A) |
A_53_P170915 |
- |
- |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) |
A_68_P05916252 |
- |
- |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) |
A_68_P05916237 |
- |
- |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) |
A_68_P05916244 |
- |
- |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P25477113 |
- |
- |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P25477109 |
- |
- |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P25477108 |
- |
- |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P25477110 |
- |
- |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P25477112 |
- |
- |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P25477111 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25477246 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25477123 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25477150 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25477193 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25477107 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25477109 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25477197 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25477235 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25477119 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25477153 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25477172 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25477209 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25477254 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25477233 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25477283 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25477141 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25477263 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25477144 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25477140 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25477223 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25477183 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25477274 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25477232 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25477203 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25477278 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25477110 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25477147 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25477169 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25477148 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25477214 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25477211 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25477154 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25477265 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25477279 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25477177 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25477113 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25477249 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25477180 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25477151 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25477256 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25477252 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25477170 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25477277 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25477239 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25477262 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25477270 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25477184 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25477220 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25477237 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25477157 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25477257 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25477114 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25477125 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25477138 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25477205 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25477261 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25477243 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25477269 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25477187 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25477158 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25477225 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25477168 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25477146 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25477199 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25477234 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25477195 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25477275 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25477210 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25477222 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25477116 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25477130 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25477155 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25477216 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25477272 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25477251 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25477181 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25477259 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25477284 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25477179 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25477111 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25477255 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25477122 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25477271 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25477212 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25477164 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25477188 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25477178 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25477115 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25477281 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25477217 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25477152 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25477190 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25477229 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25477208 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25477133 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25477161 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25477201 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25477160 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25477280 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25477241 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25477227 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25477230 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25477174 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P25477133 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P06258876 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P25477197 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P25477141 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P25477168 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P25477196 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P25477150 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P25477138 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P25477187 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P25477140 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P06547381 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P25477195 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P06066368 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P25477184 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P25477164 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P25477123 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P25477130 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P25477127 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P25477116 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P25477146 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P25477161 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P25477158 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P25477183 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P25477148 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P06547378 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P25477142 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P25477107 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P06176953 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P25477155 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P05916237 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P25477122 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P25477119 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P25477126 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P25477188 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P25477172 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P06521026 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P25477111 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P25477125 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P25477151 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P25477157 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P25477109 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P25477160 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P25477169 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P25477170 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P25477144 |
- |
- |