Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Oxr1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001130163.1 | NP_001123635.1 | 4 - 5 | 399 | 75 | |
NM_001130165.1 | NP_001123637.1 | 4 - 5 | 494 | 75 | ||
NM_001130166.1 | NP_001123638.1 | 6 - 7 | 758 | 75 | ||
NM_130885.2 | NP_570955.1 | 4 - 5 | 399 | 75 | ||
GenBank mRNA | AF324899.1 | - | 4 - 5 | 398 | 75 | |
AF333985.1 | - | 4 - 5 | 399 | 75 | ||
AK040881.1 | - | 4 - 5 | 422 | 75 | ||
AK052714.1 | - | 4 - 5 | 564 | 75 | ||
AK158703.1 | - | 6 - 7 | 740 | 75 | ||
BC089183.1 | - | 4 - 5 | 489 | 5321255 | 75 | |
BC098491.1 | - | 6 - 7 | 741 | 30086857 | 75 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons |