Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nxph1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AK047847.1 | - | 1 - 2 | 359 | 79 | |
AK083014.1 | - | 1 - 2 | 703 | 79 | ||
AK139424.1 | - | 1 - 2 | 887 | 79 | ||
AK163438.1 | - | 1 - 2 | 151 | 79 |
Set Membership: |
Amplicon length between 71 and 85 Mus musculus 1700 Probe spans exons |