Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gnai1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_010305.1 | NP_034435.1 | 1 - 2 | 366 | 64 | |
GenBank mRNA | BC032995.1 | - | 1 - 2 | 308 | 64 | |
BC138862.1 | - | 1 - 2 | 246 | 8861884 | 64 | |
BC138865.1 | - | 1 - 2 | 246 | 8861887 | 64 | |
BY740128.1 | - | 1 - 2 | 366 | 64 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus Probe spans exons |