Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Il21r | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_021887.2 | NP_068687.1 | 2 - 3 | 484 | 80 | |
GenBank mRNA | AB049137.1 | - | 2 - 3 | 376 | 80 | |
AF254068.1 | - | 2 - 3 | 188 | 80 | ||
AF269134.1 | - | 1 - 2 | 46 | 80 | ||
AF279436.1 | - | 2 - 3 | 436 | 80 | ||
AF477982.1 | - | 1 - 2 | 62 | 80 | ||
AF477983.1 | - | 1 - 2 | 62 | 80 | ||
AF477984.1 | - | 1 - 2 | 62 | 80 | ||
AF477985.1 | - | 1 - 2 | 62 | 80 | ||
AF477986.1 | - | 1 - 2 | 62 | 80 | ||
AK137793.1 | - | 2 - 3 | 451 | 80 | ||
AK150824.1 | - | 2 - 3 | 445 | 80 | ||
AK171826.1 | - | 2 - 3 | 423 | 80 | ||
AK172032.1 | - | 2 - 3 | 456 | 80 | ||
BB634399.1 | - | 2 - 3 | 484 | 80 | ||
BC057861.1 | - | 2 - 3 | 433 | 5315350 | 80 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus 1700 Probe spans exons |