Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ptpre | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_011212.3 | NP_035342.3 | 1 - 2 | 295 | 103 | |
GenBank mRNA | AK004602.1 | - | 1 - 2 | 295 | 103 | |
BC024631.1 | - | 1 - 2 | 289 | 103 | ||
D83484.1 | - | 1 - 2 | 632 | 103 | ||
U35368.1 | - | 1 - 2 | 295 | 103 | ||
U62387.1 | - | 1 - 2 | 197 | 103 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Protein Phosphatase Mammalian Gene Collection Mus musculus Probe spans exons |