Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Lat | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_010689.2 | NP_034819.1 | 9 - 10 | 863 | 122 | |
GenBank mRNA | AF036907.1 | - | 9 - 10 | 863 | 122 | |
AK088763.1 | - | 9 - 10 | 772 | 122 | ||
AK134003.1 | - | 9 - 10 | 815 | 122 | ||
BC013337.1 | - | 9 - 10 | 840 | 4191983 | 122 | |
BC052340.1 | - | 9 - 10 | 808 | 30077071 | 122 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |