Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Neurod4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_007501.4 | NP_031527.1 | 1 - 2 | 83 | 78 | |
GenBank mRNA | AK012183.1 | - | 1 - 2 | 70 | 78 | |
BC042639.1 | - | 1 - 2 | 32 | 78 | ||
BC054391.1 | - | 1 - 2 | 83 | 6491248 | 78 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons |