Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Arhgef5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_133674.1 | NP_598435.1 | 11 - 12 | 4368 | 73 | |
GenBank mRNA | AK037202.1 | - | 6 - 7 | 785 | 73 | |
BC006829.1 | - | 6 - 7 | 759 | 73 | ||
BC025127.1 | - | 6 - 7 | 655 | 73 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |