Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Grhl2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_026496.4 | NP_080772.2 | 12 - 13 | 1872 | 72 | |
GenBank mRNA | AF411213.1 | - | 12 - 13 | 1605 | 72 | |
BC004783.1 | - | 3 - 4 | 278 | 72 | ||
BC042575.1 | - | 7 - 8 | 705 | 72 | ||
BC055035.1 | - | 12 - 13 | 1661 | 6306106 | 72 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons |