Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Grhl2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_026496.4 | NP_080772.2 | 13 - 14 | 1963 | 71 | |
GenBank mRNA | AF411213.1 | - | 13 - 14 | 1696 | 71 | |
AK161180.1 | - | 6 - 7 | 641 | 71 | ||
BC004783.1 | - | 4 - 5 | 369 | 71 | ||
BC041795.1 | - | 1 - 2 | 96 | 71 | ||
BC042575.1 | - | 8 - 9 | 796 | 71 | ||
BC055035.1 | - | 13 - 14 | 1752 | 6306106 | 71 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons |