Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Copg2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_017478.3 | NP_059506.1 | 2479 | 72 | ||
GenBank mRNA | AF205065.1 | - | 2382 | 72 | ||
AJ251067.1 | - | 2451 | 72 | |||
AK147943.1 | - | 2427 | 72 | |||
BC008136.1 | - | 878 | 72 | |||
BC049178.1 | - | 2445 | 6309945 | 72 | ||
BC145898.1 | - | 2422 | 40131201 | 72 | ||
BC145900.1 | - | 2422 | 40131203 | 72 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |