Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dctn2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001190453.1 | NP_001177382.1 | 8 - 9 | 912 | 59 | |
NM_001190454.1 | NP_001177383.1 | 7 - 8 | 921 | 59 | ||
NM_027151.2 | NP_081427.1 | 7 - 8 | 906 | 59 | ||
GenBank mRNA | AK009749.1 | - | 2 - 3 | 158 | 59 | |
AK144464.1 | - | 7 - 8 | 782 | 59 | ||
AK144505.1 | - | 7 - 8 | 782 | 59 | ||
AK164021.1 | - | 7 - 8 | 783 | 59 | ||
BC004613.1 | - | 7 - 8 | 769 | 3585310 | 59 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |