Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Arhgef25 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001166413.1 | NP_001159885.1 | 9 - 10 | 959 | 106 | |
NM_028027.3 | NP_082303.2 | 9 - 10 | 1021 | 106 | ||
GenBank mRNA | AF487515.1 | - | 9 - 10 | 987 | 106 | |
AK010452.1 | - | 9 - 10 | 987 | 106 | ||
AK158684.1 | - | 9 - 10 | 959 | 106 | ||
BC041113.1 | - | 7 - 8 | 1335 | 106 | ||
BC054833.1 | - | 1 - 2 | 243 | 6439728 | 106 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |