Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Matn2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_016762.2 | NP_058042.2 | 9 - 10 | 1708 | 99 | |
GenBank mRNA | AK083151.1 | - | 9 - 10 | 1705 | 99 | |
AK138211.1 | - | 9 - 10 | 1705 | 99 | ||
AK163818.1 | - | 9 - 10 | 1667 | 99 | ||
AK209627.1 | - | 1 - 2 | 78 | 99 | ||
BC005429.1 | - | 9 - 10 | 1578 | 3492881 | 99 | |
BC092298.1 | - | 9 - 10 | 1651 | 5063535 | 99 | |
U69262.1 | - | 9 - 10 | 1707 | 99 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |